Would you like to log in again?
Human Recombinant APOJ (from E. coli)
Supplier: BioVision
AB285909-1MG AB285909-10UG AB285909-50UG
10834-778EA 972.64 USD InStock
10834-778 10834-776 10834-780
Human Recombinant APOJ (from E. coli)
Proteins and Peptides
≥9% pure active human recombinant ApoJ/Clusterin
The chaperone activity of ApoJ was confirmed by its ability to bind to proteins under physical stress such as heat (e.g. Thyroglobulin) and chemical stress such as change in pH (e.g. Procathepsin K).
Native Apolipoprotein J (ApoJ), also named Clusterin, is a heavily glycosylated, 75-8 kDa disulfide-linked heterodimeric protein. Despite being cloned since 1989, no genuine function has been attributed to ApoJ so far. The protein has been reportedly implicated in several diverse physiological processes such as sperm maturation, lipid transportation, complement inhibition, tissue remodeling, membrane recycling, cell-cell and cell-substratum interactions, stabilization of stressed proteins in a folding-competent state and promotion or inhibition of apoptosis. ApoJ gene is differentially regulated by cytokines, growth factors and stress-inducing agents. Clusterin is up- or down regulated on the mRNA or protein level in many pathological and clinically relevant situations including cancer, organ regeneration, infection, Alzheimer disease, retinitis pigmentosa, myocardial infarction, renal tubular damage, autoimmunity and others.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty