Would you like to log in again?
Human Recombinant Cystathionine ? Synthase Hr (from E. coli)
Supplier: BioVision
AB285970-500UG AB285970-100UG
10835-714EA 750.81 USD InStock
10835-714 10835-716
Human Recombinant Cystathionine ? Synthase Hr (from E. coli)
Enzymes
≥9% Pure Active Human recombinant Cystathionine β Synthase
100 U/mg
Cystathionine β-synthase (CBS; E.C. 4.2.1.22) is a PLP-dependent enzyme which plays a central role in sulfur amino acid metabolism in eukaryotes. CBS catalyzes condensation between serine and homocysteine to generate cystathionine, which is then further processed by cystathionine γ-lyase to yield cysteine. The gene encoding CBS is essentially linked to the genetic disorders of homocystinuria and Down syndrome. Homocystinuria is an autosomal recessive disease, characterized by high plasma levels of homocysteine, with clinical manifestations including mental retardation, thromboembolism and connective tissue defects. In addition, CBS also mediates synthesis of hydrogen sulfide by catalyzing condensation between cysteine and homocysteine. CBS is highly expressed in the nervous system, liver and kidney and is responsible for up to 95% of the H2S production in the brain.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty