Would you like to log in again?
Human Recombinant Ezcut Tev Protease (from E. coli)
Supplier: BioVision
AB285976-100 AB285976-1MG
10835-726EA 196.39 USD InStock
10835-726 10835-728
Human Recombinant Ezcut Tev Protease (from E. coli)
Enzymes
≥95% Pure Active Recombinant EZCut™ TEV Protease
The EZCut™ TEV Protease has an activity of ≥10,000 units/mg.
BioVision’s EZCut™ TEV Protease is a cysteine protease that recognizes the cleavage site of Glu-Xaa- Xaa-Y- Xaa-Gln-(Gly/Ser) and cleaves between Gln and Gly/Ser. The optimal sequence is Glu-Asn-Leu-Tyr-Phe-Gln-Ser/Glycine (ENLYFQS/G). It contains an enhanced form of a catalytic fragment of the NIa protein of Tobacco etch virus (TEV). TEV Protease is a restriction grade protease that has robust activity at 4C with high specificity and great stability. The optimal temperature for cleavage with this enzyme is 34°C. The protease can be used for the removal of affinity tags from fusion proteins. It contains a C-terminal His tag and can be easily removed after cleavage reactions by passing the reaction through a Ni-chelating resin. BioVision’s EZCut™ TEV Protease is an improved version of TEV protease that is highly site-specific, highly active, and significantly more stable than native TEV protease, resulting in enhanced long-term activity.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty