Would you like to log in again?
CellExp™ Human Recombinant Cathepsin D (from HEK293 cells)
Supplier: BioVision
CellExp™
AB285852-10UG
10834-406EA 781.42 USD InStock
10834-406
CellExp™ Human Recombinant Cathepsin D (from HEK293 cells)
Proteins and Peptides
≥95% pure active human cell expressed human Recombinant Cathepsin D
Cathepsin D is also known as CTSD, CPSD, which belongs to the peptidase A1 family. Cathepsin D can be cleaved into the following 2 chains: cathepsin D light chain and cathepsin D heavy chain, which is expressed in the aorta extracellular space (at protein level). The catalytic activity of Cathepsin D is specificity similar to, but narrower than, that of pepsin A. Cathepsin D does not cleave the 4-Gln-|-His-5 bond in B chain of insulin. Cathepsin D involved in the pathogenesis of several diseases such as breast cancer and possibly Alzheimer disease.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty