Would you like to log in again?
CellExp™ Human Recombinant CD55 (from HEK293 Cells)
Supplier: BioVision
CellExp™
AB285860-10UG AB285860-50UG
10834-466EA 1670.09 USD InStock
10834-466 10834-464
CellExp™ Human Recombinant CD55 (from HEK293 Cells)
Proteins and Peptides
≥95% pure active human cell expressed human Recombinant CD55/DAF
CD55, also known as DAF or decay accelerating factor, is a member of the RCA (regulators of complement activation) family characterized by four to 3 SCRs (short consensus repeats) in their plasma-exposed regions. CD55 /DAF is a 7 kDa membrane protein that regulates the complement system on the cell surface. It prevents the assembly of the C3bBb complex (the C3-convertase of the alternative pathway) or accelerates the disassembly of preformed convertase, thus blocking the formation of the membrane attack complex. This glycoprotein is broadly distributed among hematopoietic and non-hematopoietic cells. It is a determinant for the Cromer blood group system. CD55 is known to bind CD97 via the first SCR. It also binds physiologically generated C3 convertases with its second and third SCRs. Binding results in an accelerated “decay”, or dissociation of active C3 convertases, thus blocking the development of C’ attack complexes on non-foreign cells. Finally, viruses and bacteria are also known to utilize multiple SCR sites for infection.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty