Would you like to log in again?
CellExp™ Human Recombinant TFPI (from HEK293 Cells)
Supplier: BioVision
CellExp™
AB285866-10UG
10834-518EA 551.56 USD InStock
10834-518
CellExp™ Human Recombinant TFPI (from HEK293 Cells)
Proteins and Peptides
≥95% pure active human cell expressed human Recombinant TFPI
Tissue factor pathway inhibitor (TFPI) also known as Extrinsic pathway inhibitor (EPI), Lipoprotein - associated coagulation inhibitor (LACI), is a plasma proteinase inhibitor synthesized by vascular endothelial cells and part of it is associated with glycosaminoglycans of these cells. TFPI is a single-chain polypeptide which can reversibly inhibit Factor Xa (Xa) and Thrombin (Factor IIa). TFPI is a secreted protein with a N­terminal acidic region, three Kunitz (K) domains separated with by two linker regions, and a C­terminal basic region. The first K domain inhibits coagulation factor VIIa complexed to tissue factor (TF); The second K domain inhibits factor Xa; The third K domain binds to heparin; The C­terminal basic region may have several functions. For example, it plays an important role in binding of TFPI to cell surfaces.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty