Would you like to log in again?
Human Recombinant Nqo1 (from E. coli)
Supplier: BioVision
AB285901-10UG AB285901-50UG
10834-742EA 2071.28 USD InStock
10834-742 10834-740
Human Recombinant Nqo1 (from E. coli)
Proteins and Peptides
≥9% pure human Recombinant NQO1
NQO1 is a cytosolic antioxidant flavoprotein that catalyzes the reduction of highly reactive quinone metabolites and their derivatives by using NAD (P)H as an electron donor. Thus NQO1 acts as a detoxifying enzyme and is involved in the body’s protection against oxidative stress. NQO1 acts as a protein chaperone, one of its targets being p53. In humans NQO1 is expressed at high levels in adipocytes and its expression levels are positively correlated with adiposity, glucose tolerance, and makers of liver dysfunction. Altered expression of NQO1 is associated with Alzheimer’s disease. NQO1 is abnormally elevated in many types of solid tumors and may represent a useful biomarker of pancreatic cancer.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty