Would you like to log in again?
Human Recombinant PGK1 (from E. coli)
Supplier: BioVision
AB289789-100UG
10834-866EA 597.56 USD InStock
10834-866
Human Recombinant PGK1 (from E. coli)
Proteins and Peptides
≥95% Pure active Recombinant Human PGK1
Phosphoglycerate kinase 1 (PGK1) is an X-linked enzyme that plays a key role in the glycolytic pathway. The gene encoding the erythrocyte enzyme PGK1 and it seems that PGK-1 acts as a polymerase alpha cofactor protein (primer recognition protein) as a glyxolytic enzyme role. The PGK1 catalyzes the reversible conversion of 1, 3-diphosphoglycerate to 3-phosphoglycerate during glycolysis, generating one molecule of ATP. Recombinant PGK1 protein was expressed in E.coli and purified by using conventional chromatography techniques.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty