Would you like to log in again?
Human recombinant pro urokinase (from E. coli)
Supplier: BioVision
AB285954-500UG AB285954-50UG AB285954-10UG
10834-830EA 2413.18 USD InStock
10834-830 10834-828 10834-826
Human recombinant pro urokinase (from E. coli)
Enzymes
>9% Pure Recombinant human Pro-Urokinase
The enzyme is fully active as seen from its ability to cleave a fluorogenic substrate N-carbobenzyloxy-Gly-Gly-Arg-7-amido-4-methylcoumarin (Z-GGR-AMC) (Cat # K728-100).
Urokinase or Urokinase-type plasminogen activator (uPA) is a serine protease (EC 3.4.21.73). It is secreted as a single-chain zymogen, pro-Urokinase, possessing little or no intrinsic enzymatic activity. The single chain zymogen is converted into the active two chain enzyme (tcuPA) by cleavage of the bond between Lys157 and Ile158. After activation, Urokinase specifically cleaves the proenzyme plasminogen to form the active enzyme plasmin. The active plasmin then catalyzes the breakdown of fibrin polymers of blood clots. Urokinase is involved in a number of biological functions including fibrinolysis, embryogenesis, cell migration, tissue remodeling, ovulation, and wound healing. Additionally, it is a potent marker of invasion and metastasis in a variety of human cancers associated with breast, stomach, colon, bladder, ovary, brain and endometrium.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty