Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bovine Serum Albumin (BSA), Heat Shock

Supplier: BioVision
AB286866-100G AB286866-5G AB286866-25G
10835-366EA 926.98 USD InStock
10835-366 10835-368 10835-364
Bovine Serum Albumin (BSA), Heat Shock
Proteins and Peptides
SOURCE: The raw material used to manufacture this lot was derived exclusively from USDA-regulated abattoirs located in the United States. The animals received ante-mortem and post-mortem inspection under a licensed veterinarian’s supervision and were certified at the time of slaughter as free of infectious or contagious diseases and injurious parasites.

OTHER PARAMETERS:
Albumin Content: ≥ 98% by electrophoresis.
pH: 7.0 ± 0.3
Moisture: ≤ 5.0%
Heavy Metals: ≤ 20 ppm
Ash: ≤ 2%
Protease: Not Detected
DNASE: Not Detected
IgG: Not Detected
SOLUBILITY: ~ 50 mg/ml in water, with 15 min manual stirring.
APPLICATIONS/ADVANTAGES:
• Excellent in RIA and ELISA and as a diluent or blocking agent.
• No detectable protease and DNASE activity.

  • Protease-/DNASE-free

SOURCE: The raw material used to manufacture this lot was derived exclusively from USDA-regulated abattoirs located in the United States. The animals received ante-mortem and post-mortem inspection under a licensed veterinarian’s supervision and were certified at the time of slaughter as free of infectious or contagious diseases and injurious parasites.

OTHER PARAMETERS:
Albumin Content: ≥ 98% by electrophoresis.
pH: 7.0 ± 0.3
Moisture: ≤ 5.0%
Heavy Metals: ≤ 20 ppm
Ash: ≤ 2%
Protease: Not Detected
DNASE: Not Detected
IgG: Not Detected
SOLUBILITY: ~ 50 mg/ml in water, with 15 min manual stirring.
APPLICATIONS/ADVANTAGES:
• Excellent in RIA and ELISA and as a diluent or blocking agent.
• No detectable protease and DNASE activity.

SOURCE: The raw material used to manufacture this lot was derived exclusively from USDA-regulated abattoirs located in the United States. The animals received ante-mortem and post-mortem inspection under a licensed veterinarian’s supervision and were certified at the time of slaughter as free of infectious or contagious diseases and injurious parasites.

OTHER PARAMETERS:
Albumin Content: ≥ 98% by electrophoresis.
pH: 7.0 ± 0.3
Moisture: ≤ 5.0%
Heavy Metals: ≤ 20 ppm
Ash: ≤ 2%
Protease: Not Detected
DNASE: Not Detected
IgG: Not Detected
SOLUBILITY: ~ 50 mg/ml in water, with 15 min manual stirring.
APPLICATIONS/ADVANTAGES:
• Excellent in RIA and ELISA and as a diluent or blocking agent.
• No detectable protease and DNASE activity.

50 mg/ml in water, with 15 min manual stirring.
APPLICATIONS/ADVANTAGES:
• Excellent in RIA and ELISA and as a diluent or blocking agent.
• No detectable protease and DNASE activity.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty