Would you like to log in again?
Calmodulin-Sepharose Beads, BioVision Inc.
Supplier: BioVision
AB286869-1ML AB286869-10ML AB286869-5ML
10835-402EA 224.94 USD InStock
10835-402 10835-404 10835-406
Calmodulin-Sepharose Beads, BioVision Inc.
Protein Purification Protein Purification Kits
Calmodulin-Sepharose Beads are ideal for the specific enrichment of calmodulin-binding proteins from biological samples.
  • Couples recombinant human calmodulin to 6% cross-linked Sepharose beads
  • Single-step purification of native calmodulin-binding proteins
  • Ideal for the specific enrichment of calmodulin-binding proteins
Calmodulin-Sepharose Beads, ideal for the specific enrichment of calmodulin-binding proteins from biological samples
1.5 mg Calmodulin protein per ml of drained resin/beads.
Calmodulin-Sepharose Beads are prepared by covalently coupling recombinant human calmodulin (Cat # 7838) to 6% cross linked Sepharose beads. Single-step purification of native calmodulin-binding proteins. Tandem affinity purification (TAP) of protein complexes. Purification of CBP tagged proteins and calmodulin-regulated proteins from all eukaryotic cells.
Supplied as 50% aqueous slurry in 20 % Ethanol. The Ligand Density is
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty