Would you like to log in again?
Hi-Bind™, Cibacron Blue-Agarose Beads, BioVision
Supplier: BioVision
AB286867-50ML AB286867-10ML
10835-546EA 222.26 USD InStock
10835-546 10835-548
Hi-Bind™, Cibacron Blue-Agarose Beads, BioVision
Protein Purification Protein Purification Kits
Hi-Bind™ Cibacron Blue-Agarose Beads purify a variety of proteins for subsequent ELISA, immunoassays and other downstream analyses such as gel electrophoresis, functional assays and more.
  • Used as an affinity purification matrix of variety of proteins
  • Can be used for ELISA, immunoassays and other downstream analyses such as gel electrophoresis and functional assays
  • Downstream analyses
Hi-Bind™ Cibacron Blue-Agarose Beads with a binding capacity of >18 mg of Human Serum Albumin for purification of a variety of proteins for subsequent ELISA, immunoassays and other downstream analyses such as gel electrophoresis, functional assays and more.
Formulation is provided as 50% aqueous slurry containing 20% ethanol. The Ligand Density is 8-10 µmol Cibacron Blue 3G-A per ml of drained gel/resin. The binding capacity of the Hi-Bind™ Cibacron Blue 3G-A-Agarose beads is >18 mg of Human Serum Albumin per ml of the drained gel/resin.
BioVision’s Hi-Bind™ Cibacron Blue-Agarose beads can be used as an affinity purification matrix of variety of proteins for subsequent ELISA, immunoassays and other downstream analyses such as gel electrophoresis, functional assays, and more.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty