Would you like to log in again?
Protein G Coated 96-Well Plates, BioVision Inc.
Supplier: BioVision
AB286902-1UNITS
10835-318EA 108.55 USD InStock
10835-318
Protein G Coated 96-Well Plates, BioVision Inc.
Microplates Cell Culture Plates
Protein G coated plates for detecting low amounts of antibodies using labeled antigens.
  • Detects low amounts of antibodies
  • Increase assay sensitivity
  • Labeled antigens used for detection
Protein G coated plates for detecting low amounts of antibodies using labeled antigens. These pre-coated plates can increase assay sensitivity and are ideal when a low amount of antibody is available.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty