Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Goat Antibody (Alexa Fluor® 594)

Supplier: Jackson Immunoresearch Laboratories
112-585-175
102645-958EA 291.69 USD InStock
102645-958
Anti-IgG Goat Antibody (Alexa Fluor® 594)
Antibodies
Whole IgG antibodies are isolated as intact molecules from antisera by immunoaffinity chromatography. They have an Fc portion and two antigen binding Fab portions joined together by disulfide bonds and therefore they are divalent. The average molecular weight is reported to be about 160 kDa. The whole IgG form of antibodies is suitable for the majority of immunodetection procedures and is the most cost effective. Based on antigen-binding assay, Western blotting, and/or ELISA, the antibody reacts with the light chains on rat IgG and with those common to other rat immunoglobulins. Reaction is primarily with kappa light chains. The antibody does not react with the heavy chain of rat IgG. The antibody has been tested by ELISA to ensure minimal cross-reaction with bovine, goat, horse, human, mouse, rabbit and sheep immunoglobulins, but it may cross-react with immunoglobulins from other species.

Type:
Antigen: IgG light chain
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 594
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty