Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Bovine Antibody (Alexa Fluor® 594)

Supplier: Jackson Immunoresearch Laboratories
805-585-180
102650-222EA 209.23 USD InStock
102650-222
Anti-IgG Bovine Antibody (Alexa Fluor® 594)
Antibodies
Whole IgG antibodies are isolated as intact molecules from antisera by immunoaffinity chromatography. They have an Fc portion and two antigen binding Fab portions joined together by disulfide bonds and therefore they are divalent. The average molecular weight is reported to be about 160 kDa. The whole IgG form of antibodies is suitable for the majority of immunodetection procedures and is the most cost effective. Based on immunoelectrophoresis and/or ELISA, the antibody reacts with whole molecule goat IgG. It also reacts with the light chains of other goat immunoglobulins. No antibody was detected against non-immunoglobulin serum proteins. The antibody has been tested by ELISA and/or solid-phase adsorbed to ensure minimal cross-reaction with bovine, chicken, guinea pig, syrian hamster, horse, human, mouse, rabbit and rat serum proteins, but it may cross-react with immunoglobulins from other species.

Type:
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 594
Epitope:
Host: Bovine
Isotype: IgG
Reactivity: Goat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty