Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG gamma Goat Polyclonal Antibody (CF™ 543)

Supplier: Biotium

20445-50UL 20445-500UL
10001-884EA 103.9 USD InStock
10001-884 10001-882
Anti-IgG gamma Goat Polyclonal Antibody (CF™ 543)
Antibodies
Goat anti-human IgG, Fc gamma specific antibody reacts specifically with the human IgG heavy (γ) chains and not with light chains and F(ab’)2 fragment on all human immunoglobulins. To minimize cross-reactivity, it has been adsorbed against bovine, horse, mouse and rabbit serum proteins prior to conjugation. CF™543 is an orange fluorescent dye with an excitation spectrum optimally matching the 543 laser line. Antibody conjugates of the dye are much brighter than those of Alexa Fluor® 546.

Alexa Fluor® is a registered trademark of Thermo Fisher Scientific.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF543
Epitope:
Host: Goat
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty