Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Fcgamma Goat Polyclonal Antibody (CF™ 640R)

Supplier: Biotium

20410-500UG
10156-348EA 208.83 USD InStock
10156-348
Anti-IgG Fcgamma Goat Polyclonal Antibody (CF™ 640R)
Antibodies
This product is prepared by labeling F(ab’)2 fragment goat anti-mouse IgG (H+L), Fcγ fragment specific, with CF™640R. The antibody has been adsorbed against human, bovine and horse serum proteins to minimize cross-reactivity. Far-red fluorescent CF™640R (Ex/Em 642/662 nm) is much brighter than Cy®5 and at least as bright as Alexa Fluor® 647, and has exceptional photostability.

Alexa Fluor® is a registered trademark of Thermo Fisher Scientific. Cy®5 is a registered trademark of GE Healthcare.

Type: Secondary
Antigen: IgG Fcgamma
Clonality: Polyclonal
Clone:
Conjugation: CF640R
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty