Would you like to log in again?
PEEK Tubing, PerkinElmer
Supplier: Perkin Elmer
N9302650
45600-118EA 130.26 USD InStock
45600-118
PEEK Tubing, PerkinElmer
Tubing Chromatography Tubing
PEEK tubing has the strength required to withstand continuous use at HPLC pressure without swelling or bursting.
  • Can be used virtually with any organic or inorganic liquid
  • Not affected by halide salts, high-strength buffers, or other aggressive mobile phases
PEEK tubing acts as an excellent altnerative for stainless steel tubing in high-pressure applications and analytical systems. Its mechanical properties allow it to be used in many situations where stainless steel would become too reactive.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty