Would you like to log in again?
AlphaTec™ 56-016 PVC Apron, Ansell
Supplier: Ansell
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
AlphaTec®
950094
35555-888CS 326.32 USD InStock
35555-888
AlphaTec™ 56-016 PVC Apron, Ansell
Disposable Garments
This medium-duty apron features 5- to 6-millimeter die-cut PVC and offers users a combination of chemical splash protection and flexible comfort.
  • Seamless construction offers resistance to puncture and abrasions
  • Superior flexibility
  • Adjustable ties ensure optimal fit
The one-piece construction of these aprons is economical, and they feature integral head openings and ties
Use in the food processing, food service, and light manufacturing industries. Uses include food handling and preparation, industrial maintenance, and chemical processing.
Seamless vinyl construction minimizes opportunity to trap contaminants and offers resistance to puncture and abrasions.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty