Would you like to log in again?
mySPIN™ Mini Centrifuges Series, Thermo Scientific
Supplier: Thermo Scientific
mySPIN™
75004061 75004081
10199-454EA 1764.04 USD InStock
10199-454 10199-452
mySPIN™ Mini Centrifuges Series, Thermo Scientific
Centrifuges Microcentrifuges
Bring increased efficiency and user comfort to your lab with mySPIN™ 6 and 12 mini centrifuges.
  • Quiet operation, low vibration
  • Ability to perform fast spins with higher RCF requirements
  • Palm-shaped lid for left- or right-handed users
  • mySPIN 12 centrifuge is ideal for most protocols requiring fast spins and appropriate for PCR, microfilter cell separation, and HPLC protocols
  • mySPIN 6 is ideal for microfilter cell separations and HPLC samples, with adapters to accommodate all major microtube and PCR tube styles (2.0, 1.5, 0.5, 0.2 mL)
Tool-free rotors and quick-spin options make them fast and easy to use. Benefit from a compact footprint that fits into your workstation and ergnonomic design that stands out. Ideal for microfilter cell separation and HPLC protocols, with the mySPIN 12 centrifuge also supporting PCR, as well as clinical and diagnostic applications.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty