Would you like to log in again?
HyFlex® 11-200 Cut-Resistant Sleeve, Ansell
Supplier: Ansell
HyFlex®
284388
10049-080CS 3054.33 USD InStock
10049-080
HyFlex® 11-200 Cut-Resistant Sleeve, Ansell
Workwear
This high-visibility sleeve delivers comfortable fit through its anatomical design with an adjustable system in the forearm area and generous space in the bicep area to fit all wearers.
  • Sleeves are resistant to cuts
  • Generous space allows freedom of movement for all wearers
  • Comfortable fit
  • Ambidextrous
The 11-200 delivers increased safety for the worker, and increased productivity and worker acceptance through freedom of movement.
Use in the aerospace, automotive manufacturers and OEM, electrical machinery, glass, machinery, metal fabrication, transport equipment, and white goods industries. Uses include body shop (excluding welding), glass sheet handling, metal sheet handling, and stamping.
Certifications: ANSI Level 3 Cut Protection; ANSI Level 2 Abrasion Protection; EN 388:2542
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty