Would you like to log in again?
Albumin Reagent Set, Pointe Scientific
Supplier: Pointe Scientific
OA902-792
10023-456EA 505.26 USD InStock
10023-456
Albumin Reagent Set, Pointe Scientific
Assays Protein Assays
The Albumin Reagent Set is used for the quantitative determination of Albumin in serum.
  • Quantitative determination of Albumin in serum
  • Increase in blue-green color
  • Standard is used to calibrate the determination of unknown specimens
Used for the quantitative determination of Albumin in serum
The Olympus Albumin Reagent is used on the Beckman Coulter AU 400, 480, and 640 analyzers and is for in vitro diagnostic use only. This reagent is measured at 600/700nm.
Albumin is bound by the BCG dye to produce an increase in the blue-green color measured at 630 nm. The color increase is proportional to the concentration of albumin present.
Certifications: CE Compliant
Ordering information: Materials Required but not Provided: Either Chemistry Calibrator or Albumin Standard; as well as both Normal and Abnormal Control Sera with known Albumin Concentrations Required.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty