Would you like to log in again?
Depyrogenated Glass Vials, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
23-CTP4/PF
10142-538CS 760.05 USD InStock
10142-538
Depyrogenated Glass Vials, Thermo Scientific
Vials
Save on capital equipment with ready-to-use depyrogenated glassware in a wide variety of sizes and configurations.
Cleaning and packaging is performed in a Class 100/10 Cleanroom
Recommended for endotoxin-free application
Certifications: Certificate of Analysis (COA) is included with each lot.
Ordering information: Containers with additional low particle certification are available. Thermo Scientific also offers glass vials specially prepared to meet endotoxin levels of less than 0.06 EU/ml. Ask your customer service representative about our depyrogenation service with our validated process, performed in our Class 100 Cleanroom.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty