Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Uridine 99%

Supplier: Thermo Scientific Chemicals
Synonyms: 1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dioneUracil-1-β-D-ribofuranoside1-β-D-Ribofuranosyluracil

AC14077-1000 AC14077-0010 AC14077-0250
200000-996EA 127.11 USD InStock
200000-996 200008-046 103516-478
Uridine 99%
Uridine


Formula: C₉H₁₂N₂O₆
MW: 244.20 g/mol
Melting Pt: 162…170 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006526
CAS Number: 58-96-8
EINECS: 200-407-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty