Would you like to log in again?
HotStart and WarmStart Enzyme Modification Kits, Biotium
Supplier: Biotium
29053-T 29053 29054
89493-570EA 310.61 USD InStock
89493-570 89493-040 10018-154
HotStart and WarmStart Enzyme Modification Kits, Biotium
Assays Enzyme Assays
Reversibly modify lysine residues on enzymes, rendering them inactive. Reverse the modification by heating.
  • Use HotStart Polymerase Modification Kit to make inactive thermostable DNA polymerase that can be activated by heating to >90 °C
  • Use WarmStart™ Enzyme Modification Kit to inactivate non-thermostable enzymes like reverse transcriptase
  • Restore protein activity by heating to 45 °C
The HotStart Polymerase Modification Kit provides an easy way to reversibly modify lysine residues of thermostable DNA polymerase, rendering the enzyme inactive
The WarmStart™ Enzyme Modification Kit allows temperature control of enzyme activity to be applied to non-thermostable enzymes like reverse transcriptase. WarmStart™ protein activity is restored by heating to 45 °C or higher. The trial size WarmStart™ modification kit contains reagents for modifying up to 0.1 mg protein. The full size kit contains reagents for modifying up to 0.5 mg protein.
The modification is reversed after heating to >90 °C. HotStart modification of DNA polymerase for PCR prevents amplification of non-specific PCR products due to low stringency annealing of primers at low temperature during reaction assembly. The regular size kit is sufficient for modifying 0.5 mg polymerase.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty