Would you like to log in again?
Precision™ High-Performance Ovens, Thermo Scientific
Supplier: Thermo Scientific
Precision™
6052 6054 6056 6051 6055 6053 6050
13304-216EA 12886.53 USD InStock
13304-216 52352-124 52352-118 52352-116 13304-218 52352-122 52352-120
Precision™ High-Performance Ovens, Thermo Scientific
Ovens Standard Ovens
Conduct drying applications requiring high temperatures, ultra-precise temperature control and a very broad temperature range of ambient +15° to 325°C stability with Thermo Scientific™ Precision™ High-Performance Ovens.
  • Bright LED display
  • All stainless-steel interior
  • Stainless-steel exterior door with fiberglass gasket for secure, tight seal prevents heat loss
  • Highly responsive RTD sensor provides temperature sensitivity of ±0.1°C or better
  • Low-density heating elements ensure long life
  • Turbo blowers for efficient drying are side-mounted within stainless-steel chamber
These ovens feature high-quality construction and high-efficiency insulation, providing temperature uniformity as precise as ±0.5°C with excellent stability and air exchange within the chamber. The built-in 12-hour mechanical timer along with user-adjustable safety thermostat allows for power cut-out in case of over temperature. Nontip shelves spaced 4cm (1.6") apart quickly, easily remove for thorough cleaning.
Certifications: ASTM E145 Type IIA, ASTM D-2436, and UL 746B performance/uniformity standards. UL listed.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty