Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Staurosporine

Supplier: Biotium

Synonyms: Antibiotic AM-2282(5S,6R,7R,9R)-6-Methoxy-5-methyl-7-(methylamino)-6,7,8,9,15,16-hexahydro-5H,14H-17-oxa-4b,9a,15-triaza-5,9-methanodibenzo[b,h]cyclonona[jkl]cyclopenta[e]-as-indacen-14-one

00025
89138-114EA 130.6 USD InStock
89138-114
Staurosporine
Staurosporine
Staurosporine is a potent apoptosis inducer.

  • Potent and cell-permeable inhibitor
  • Induces apoptosis in human neuroblastoma cells
  • Apoptosis in chick embryonic neurons

Staurosporine is a potent and cell-permeable inhibitor of a wide variety of protein kinases including protein kinase C (IC50 = 0.7 nM), protein kinase A (IC50 = 7 nM), and protein kinase G (IC50 = 8.5 nM). Staurosporine induces apoptosis in human neuroblastoma cells and chick embryonic neurons.


Formula: C₂₈H₂₆N₄O₃
MW: 466.541 g/mol
MDL Number: MFCD18252446
CAS Number: 62996-74-1
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty