Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Orion™ Triode™ 3-in-1 pH/ATC Probe, Thermo Scientific

Supplier: Thermo Scientific
Orion™
9109WP 9109WL
21000-020EA 808.86 USD InStock
21000-020 34104-186
Orion™ Triode™ 3-in-1 pH/ATC Probe, Thermo Scientific
Electrodes pH Electrodes
Operate the highly-versatile Orion™ Triode™ 3-in-1 pH/ATC Probe in either the field or a plant.

  • pH range of 0 to 14
  • Ag/AgCl sealed internal reference system
  • Wick junction

Contains a low-maintenance, gel-filled electrode with built-in ATC.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty