Would you like to log in again?
Human Recombinant Carbonic Anhydrase 7 (from E. coli)
Supplier: ProSci
91-094
75788-912EA 533.61 USD InStock
75788-912
Human Recombinant Carbonic Anhydrase 7 (from E. coli)
Enzymes
Carbonic Anhydrase 7 (CA7) is a member of the alpha-carbonic anhydrase family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Alpha-carbonic anhydrase is a large family of zinc metalloenzymes that catalyze the reversible hydration of carbon dioxide. Furthermore, Alpha-carbonic anhydrase is associated with many biological processes, including calcification, respiration, bone resorption, acid-base balance and the formation of aqueous humor. CA7 is activated by histamine, L-adrenaline, L- and D-histidine, and L- and D-phenylalanine, but it is inhibited coumarins, sulfonamide derivatives such as acetazolamide (AZA) by saccharin and Foscarnet.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty