Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Human Recombinant N-Glycosylase (from E. coli)
Supplier: ProSci
91-111
75788-938EA 700.27 USD InStock
75788-938
Human Recombinant N-Glycosylase (from E. coli)
Enzymes
Human N-Glycosylase/DNA Lyase is a DNA repair enzyme, which incises DNA at 8-oxoG residues, and excises 7,8-dihydro-8-oxoguanine and 2,6-diamino-4-hydroxy-5-N-methylformamidopyrimidine (FAPY) from damage DNA
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-GST tag
DGG1 has a beta -lyase activity that nicks DNA 3’ to the lesion.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty