Would you like to log in again?
Human Recombinant Flap Endonuclease 1 (from E. coli)
Supplier: ProSci
91-123
75788-962EA 700.27 USD InStock
75788-962
Human Recombinant Flap Endonuclease 1 (from E. coli)
Enzymes
Flap Endonuclease 1 (FEN1) is a member of the XPG/RAD2 endonuclease family
This recombinant protein can be used for biological assays. For research use only.
During DNA replication, FEN1 cleaves the 5'-overhanging flap structure and processes the 5' ends of Okazaki fragments for synthesis. FEN1 also exhibits RNase H activity by possessing 5'-3' exonuclease activity on gapped double-stranded or nicked DNA, FEN1 is involved in the long patch base excision repair (LP-BER) pathway, it can cleave within the apurinic/apyrimidinic (AP) site-terminated flap. FEN1 can prevent flaps from equilibrating into structures that lead to duplications and deletions. FEN1 is also involved in replication and repair of rDNA and in repairing mitochondrial DNA.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty