Would you like to log in again?
Human Recombinant Cyclin-Dependent Kinase 2 (from E. coli)
Supplier: ProSci
91-191
75789-098EA 946.47 USD InStock
75789-098
Human Recombinant Cyclin-Dependent Kinase 2 (from E. coli)
Enzymes
Cyclin-dependent kinase 2 (CDK2) belongs to the cyclin-dependent kinase of Ser/Thr protein kinase
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
CDK2 acts as a catalytic subunit of the cyclin dependent kinase complex, whose activity is restricted to the G1-S phage of the cell cycle, it is essential for the G1/S transition. The kinase activity of CDK2 can be regulated by the association with a cyclin subunit, its phosphorylation state and CDK inhibitors. The activation of the CDK2/cyclin complex requires the phosphorylation of Thr160 and the dephosphorylation of Try14 and Tyr15. The inhibition of CDK2-cyclin complex can also be attributed to association with p27Kip1 and p21Waf1/Cip1. The activation of CDK2 has been shown to be necessary for apoptosis of quiescent cells, such as neurons, thymocytes and endothelial cells.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty