Would you like to log in again?
Human Recombinant Malate dehydrogenase (MDH) (from E. coli)
Supplier: ProSci
91-256
75789-188EA 946.47 USD InStock
75789-188
Human Recombinant Malate dehydrogenase (MDH) (from E. coli)
Enzymes
Malate Dehydrogenase, Cytoplasmic (MDH1) is an enzyme which belongs to the MDH Type 2 sub-family of LDH/MDH superfamily
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
MDH1 is involved in the Citric Acid Cycle that catalyzes the conversion of Malate into Oxaloacetate (using NAD+) and vice versa. MDH1 should not be confused with Malic Enzyme, which catalyzes the conversion of Malate to Pyruvate, producing NADPH. MDH1 also participates in Gluconeogenesis, the synthesis of Glucose from smaller molecules. Pyruvate in the mitochondria is acted upon by Pyruvate Carboxylase to form Pxaloacetate, a Citric Acid Cycle intermediate. In order to transport the Oxaloacetate out of the Mitochondria, Malate Dehydrogenase reduces it to Malate, and it then traverses the inner Mitochondrial membrane. Once in the cytosol, the Malate is oxidized back to Oxaloacetate by MDH1. Finally, Phosphoenol-Pyruvate Carboxy Kinase (PEPCK) converts Oxaloacetate to Phosphoenol Pyruvate.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty