Would you like to log in again?
Human Recombinant Carbonic Anhydrase 10 (from Cells)
Supplier: ProSci
91-285
75789-246EA 912.46 USD InStock
75789-246
Human Recombinant Carbonic Anhydrase 10 (from Cells)
Enzymes
Carbonic Anhydrase X (CA10) belongs to CA family of zinc metalloenzymes, which catalyze the reversible hydration of carbon dioxide in various biological processes such as respiration, renal tubular acidification and bone resorption
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
While CA10 is a secreted protein without Carbonic Anhydrase activity (i.e., the reversible hydration of CO2) due to point mutations in the zinc binding site, it has esterase activity. The human and mouse CA10 are expressed in the brain, indicating that they may play a role in brain development.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty