Would you like to log in again?
Human Recombinant alpha-Galactosidase A (from Cells)
Supplier: ProSci
91-323
75789-322EA 785.03 USD InStock
75789-322
Human Recombinant alpha-Galactosidase A (from Cells)
Enzymes
alpha-Galactosidase A is a homodimeric glycoprotein that belongs to the glycosyl hydrolase 27 family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
It is a lysosomal enzyme and used as a long-term enzyme replacement therapy in patients with a confirmed diagnosis of Fabry disease. alpha-Galactosidase A can hydrolyze terminal alpha-galactosyl moieties from glycolipids and glycoproteins and catalyze the hydrolysis of melibiose into galactose and glucose. Defects alpha-Galactosidase A are the cause of Fabry disease (FD) which is a rare X-linked sphingolipidosis disease with glycolipid accumulates in many tissues. The disease consists of an inborn error of glycosphingolipid catabolism. FD patients show systemic accumulation of globotriaoslyceramide (Gb3) and related glycosphingolipids in the plasma and cellular lysosomes throughout the body. Patients may show ocular deposits, febrile episodes, and burning pain in the extremities. Death results from renal failure, cardiac or cerebral complications of hypertension or other vascular disease.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty