Would you like to log in again?
Human Recombinant N-Acetylglucosamine-6-Sulfatase (from Cells)
Supplier: ProSci
91-325
75789-326EA 953.38 USD InStock
75789-326
Human Recombinant N-Acetylglucosamine-6-Sulfatase (from Cells)
Enzymes
N-Acetylglucosamine-6-Sulfatase is a member of the Sulfatase family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
N-Acetylglucosamine-6-Sulfatase is required for the lysosomal degradation of the Glycosaminoglycans (GAG) Heparan Sulfate and Keratan Sulfate. N-Acetylglucosamine-6-Sulfatase hydrolyzes the 6-Sulfate groups of the N-Acetyl-D-Glucosamine 6-Sulfate units of Heparan Sulfate and Keratan Sulfate. N-Acetylglucosamine-6-Sulfatase binds 1 Calcium ion per subunit. N-Acetylglucosamine-6-Sulfatase deficiency are the cause of Mucopolysaccharidosis Type 3D (MPS3D), an inborn error leading to lysosomal accumulation of heparan sulfate. MPS3D has profound mental deterioration, hyperactivity, and relatively mild somatic manifestations.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty