Would you like to log in again?
Human Recombinant Carnosine Dipeptidase 1 (from Cells)
Supplier: ProSci
91-392
75789-460EA 946.47 USD InStock
75789-460
Human Recombinant Carnosine Dipeptidase 1 (from Cells)
Enzymes
Carnosine Dipeptidase 1 (CNDP1) belongs to the M20 metalloprotease family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
CNDP1 is specifically expressed in the brain, serum and adult nervous central system. It is identified as human carnosinase. CNDP1 contains trinucleotide (CTG) repeat length polymorphism in the coding region and is inhibited by the metal chelator 1,10-o-phenantrolin. In addition, CNDP1 can hydrolyse the beta-Ala|-His dipeptide (carnosine), anserine, Xaa-|-His dipeptides and other dipeptides including homocarnosine. CNDP1 deficiency has been associated with homocarnosinosis disease.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty