Would you like to log in again?
Human Recombinant Chitinase-3-Like 1 (from Cells)
Supplier: ProSci
91-408
75789-492EA 623.74 USD InStock
75789-492
Human Recombinant Chitinase-3-Like 1 (from Cells)
Enzymes
Chitinase-3-Like Protein 1 (CHI3L1) belongs to the glycosyl hydrolase 18 family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
CHI3L1 is expressed in activated macrophages, articular chondrocytes, synovial cells as well as in liver. It lacks chitinase activity and is secreted by activated macrophages, chondrocytes, neutrophils and synovial cells. CHI3L1 is thought to play a role in defense against pathogens, or in tissue remodeling, and in the capacity of cells to respond to and cope with changes in their environment. In addition, CHI3L1 is associated with susceptibility to asthma-related traits type 7 (ASRT7) which assessed by methacholine challenge test, serum IgE levels, atopy, and atopic dermatitis.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty