Would you like to log in again?
Human Recombinant Chymotrypsin-Like Protease (from Cells)
Supplier: ProSci
91-414
75789-504EA 700.27 USD InStock
75789-504
Human Recombinant Chymotrypsin-Like Protease (from Cells)
Enzymes
Chymotrypsin-Like Protease CTRL-1 is a protease that belongs to the peptidase S1 family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Human CTRL-1 is synthesized as a 264 amino acid (aa) precursor that contains an 18 aa signal sequence, 15 aa activation peptide and a 231 aa mature chain. CTRL-1 Contains 1 peptidase S1 domain. It has many molecular functions, such as hydrolase, protease, and serine protease. CTRL-1 plays a role in digest and hydrolyze proteins in biological process.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty