Would you like to log in again?
Human Recombinant beta-1,4-Galactosyltransferase 4 (from Cells)
Supplier: ProSci
91-464
75789-604EA 700.27 USD InStock
75789-604
Human Recombinant beta-1,4-Galactosyltransferase 4 (from Cells)
Enzymes
beta -1,4-galactosyltransferase 4 (B4GALT4) is a single-pass type II membrane protein that belongs to the Glycosyltransferase 7 family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
B4GALT4 consist of the following 2 domains: N-Acetyllactosamine Synthase and beta -N-Acetylglucosaminyl-Glycolipid beta -1,4-Galactosyltransferase. B4GALT4 is highly expressed in the heart, placenta, kidney, and pancreas; it is lowly expressed in the brain, colon, lung, muscle, ovary, testis, and uterus. B4GALT4 function is responsible for the synthesis of complex-type N-linked oligosaccharides in many glycoproteins as well as the carbohydrate moieties of glycolipids.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty