Would you like to log in again?
Human Recombinant Butyrylcholine Esterase (from Cells)
Supplier: ProSci
91-508
75789-692EA 946.47 USD InStock
75789-692
Human Recombinant Butyrylcholine Esterase (from Cells)
Enzymes
Butyrylcholine Esterase (BCHE) is a secreted protein that belongs to the type-B carboxylesterase/lipase family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
BCHE is a major acetylcholine hydrolyzing enzyme in the circulation. It is detected in blood plasma and present in most cells except erythrocytes. BCHE is an esterase with broad substrate specificity. BCHE can contribute to the inactivation of the neurotransmitter acetylcholine. BCHE can degrade a large number of neurotoxic organophosphate esters. Thus, it plays important pharmacological and toxicological roles and is thought to be involved in the pathological progression. Defects in BCHE are the cause of butyrylcholinesterase deficiency (BChE deficiency) which is a metabolic disorder characterized by prolonged apnoea after the use of certain anesthetic drugs, including the muscle relaxants succinylcholine and other ester local anesthetics.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty