Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Human Recombinant Ribonuclease 3 (from Cells)
Supplier: ProSci
91-718
75790-096EA 946.47 USD InStock
75790-096
Human Recombinant Ribonuclease 3 (from Cells)
Enzymes
Ribonuclease 3 (RNASE3) is a basic protein that is localized to the eosinophil primary matrix and belongs to the pancreatic ribonuclease family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
RNASE3 is released during degranulation of eosinophils. RNASE3 possesses a wide variety of biological activities. RNASE3 interacts with bacterial lipopolysaccharide (LPS) and lipoteichoic acid (LTA). RNASE3 exhibits antibacterial activity, including cytoplasmic membrane depolarization of preferentially Gram-negative, but also Gram-positive strains. It promotes E. coli outer membrane detachment, alteration of the overall cell shape and partial loss of cell content.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty