Would you like to log in again?
Human Recombinant Sulfamidase (from Cells)
Supplier: ProSci
91-726
75790-112EA 946.47 USD InStock
75790-112
Human Recombinant Sulfamidase (from Cells)
Enzymes
N-Sulphoglucosamine Sulphohydrolase (SGSH) is an important member of the sulfatase family which is involved in the degradation of heparin sulfate
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
SGSH binds one calcium ion per subunit as a cofactor. SGSH catalyzes N-sulfo-D-glucosamine and H2O to D-glucosamine and sulfate. SGSH deficiency is result in mucopolysaccharidosis type 3A (MPS3A), a recessive lysosomal storage disease characterized by neurological dysfunction but relatively mild somatic manifestations.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty