Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Human Recombinant Proprotein Convertase 9 (from Cells)
Supplier: ProSci
91-785
75790-222EA 700.27 USD InStock
75790-222
Human Recombinant Proprotein Convertase 9 (from Cells)
Enzymes
Recombinant Human Proprotein Convertase Subtilisin/Kexin Type 9/PCSK9 (D374Y) is a gain of function mutant of human PCSK9 protein
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Human PCSK9 is a secretory subtilase belonging to the proteinase K subfamily. PCSK9 is synthesized as a soluble zymogen that undergoes autocatalytic intramolecular processing in the ER, the pro domain and mature chain are secreted together through noncovalent interactions. PCSK9 binds with low-density lipoprotein receptor (LDLR) and it plays a major regulatory role in cholesterol homeostasis. Inhibition of PCSK9 function by preventing PCSK9/LDLR interaction is currently being explored as a means of lowering cholesterol levels. PCSK9 also binds to apolipoprotein receptor 2 (ApoER2), and play a role in the neural development.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty