Would you like to log in again?
Human Recombinant Palmitoyl-Protein Thioesterase 1 (from Cells)
Supplier: ProSci
91-846
75788-638EA 629.48 USD InStock
75788-638
Human Recombinant Palmitoyl-Protein Thioesterase 1 (from Cells)
Enzymes
Palmitoyl-protein thioesterase 1(PPT-1 for short), also known as Palmitoyl-protein hydrolase 1, belongs to the palmitoyl-protein thioesterase family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
It is a small glycoprotein involved in the catabolism of lipid-modified proteins during lysosomal degradation. This enzyme removes thioester-linked fatty acyl groups such as palmitate from modified cysteine residues in proteins or peptides during lysosomal degradation. Defects in PPT1 are the cause of neuronal ceroid lipofuscinosis type 1.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty