Would you like to log in again?
Human Recombinant beta-1,4-Galactosyltransferase 3 (from Cells)
Supplier: ProSci
91-863
75790-362EA 946.47 USD InStock
75790-362
Human Recombinant beta-1,4-Galactosyltransferase 3 (from Cells)
Enzymes
Beta-1,4-galactosyltransferase 3 (B4GALT3) belongs to the glycosyltransferase 7 family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
It is responsible for the synthesis of complex-type N-linked oligosaccharides in many glycoproteins as well as the carbohydrate moieties of glycolipids. It is highest expression in placenta, prostate, testis, ovary, intestine and muscle, and in fetal brain.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty