Would you like to log in again?
Human Recombinant ASM-like phosphodiesterase 3a (from Cells)
Supplier: ProSci
91-932
75790-500EA 912.46 USD InStock
75790-500
Human Recombinant ASM-like phosphodiesterase 3a (from Cells)
Enzymes
Acid sphingomyelinase-like phosphodiesterase 3a (SMPDL3A ) is a novel liver X receptor (LXR) -regulated gene, with an LXR response element within its promoter
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
The induction of SMPDL3A is LXR-dependent and is restricted to human blood cells with no induction observed in mouse cellular systems. LXR function as physiological sensors of cholesterol metabolites (oxysterols), regulating key genes involved in cholesterol and lipid metabolism. LXRs have been extensively studied in both human and rodent cell systems, revealing their potential therapeutic value in the contexts of atherosclerosis and inflammatory diseases.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty