Would you like to log in again?
Human Recombinant GDP-L-Fucose Synthase (from E. coli)
Supplier: ProSci
91-969
75790-574EA 946.47 USD InStock
75790-574
Human Recombinant GDP-L-Fucose Synthase (from E. coli)
Enzymes
GDP-L-Fucose Synthase is a NADP(H)-binding protein
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
It catalyzes the two-step epimerase and the reductase reactions in GDP-D-mannose metabolism, converting GDP-4-keto-6-D-dexoymannose to GDP-L-fucose. GDP-L-Fucose is the substrate of several fucosyltransferase, involving the expression of mamy glycoconjugates, including blood group ABH antigens and development adhesion antigens. Mutations in the TSTA3 gene may cause leukocyte adhesion deficiency type II.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty