Would you like to log in again?
Human Recombinant Cysteine Dioxygenase Type 1 (from E. coli)
Supplier: ProSci
91-984
75790-604EA 464.46 USD InStock
75790-604
Human Recombinant Cysteine Dioxygenase Type 1 (from E. coli)
Enzymes
Cysteine Dioxygenase Type 1 (CDO1) is a mammalian non-heme iron enzyme that belongs to the cysteine dioxygenase family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
CDO1 is highly expressed in the liver and placenta, and has a low expression in heart, brain and pancreas. CDO1 can also be detected in hepatoblastoma HepG2 cells. CDO1 catalyzes the conversion of L-cysteine to cysteine sulfinic acid by incorporation of dioxygen. CDO1 is a vital regulator of cellular cysteine concentrations and has an essential role in maintaining the hepatic concentration of intracellular free cysteine within a proper narrow range. CDO1 is able to alter intracellular cysteine levels and glutathione levels.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty