Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Human Recombinant Argininosuccinate Synthase (from E. coli)
Supplier: ProSci
91-986
75790-608EA 946.47 USD InStock
75790-608
Human Recombinant Argininosuccinate Synthase (from E. coli)
Enzymes
Argininosuccinate Synthase (ASS1) is an urea cycle enzyme with a tetrameric structure composed of identical subunits
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
ASS1 is involved in the synthesis of arginine and catalyzes that condensation of citrulline and aspartate to argininosuccinate using ATP. ASS1 is important to the urea cycle as it catalyzes the important second last step in the arginine biosynthetic pathway. ASS1 mainly expressed in periportal hepatocytes, but also in most other body tissues. A deficiency of ASS1 causes citrullinemia (CTLN1), an autosomal recessive disease which is characterized by severe vomiting spells and mental retardation.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty